Skip to Content
Merck
All Photos(1)

Key Documents

3542

Sigma-Aldrich

Glutathione, Oxidized, Free Acid

Glutathione, Oxidized, Free Acid, CAS 27025-41-8, forms a "redox buffer" in physiological systems in the presence of reduced glutathione.

Synonym(s):

Glutathione, Oxidized, Free Acid, Cyclic(γ-Glu-Cys-Gly)₂, GSSG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H32N6O12S2
CAS Number:
Molecular Weight:
612.63
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.25
Pricing and availability is not currently available.

Quality Level

description

RTECS - MC0556500
Merck USA index - 14, 4475

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water: 1 mg/mL
DMF: 10 mg/mL
ethanol: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

S(SCC(NC(=O)CCC(N)C(=O)O)C(=O)NCC(=O)O)CC(NC(=O)CCC(N)C(=O)O)C(=O)NCC(=O)O

InChI

1S/C20H32N6O12S2/c21-9(19(35)36)1-3-13(27)25-11(17(33)23-5-15(29)30)7-39-40-8-12(18(34)24-6-16(31)32)26-14(28)4-2-10(22)20(37)38/h9-12H,1-8,21-22H2,(H,23,33)(H,24,34)(H,25,27)(H,26,28)(H,29,30)(H,31,32)(H,35,36)(H,37,38)

InChI key

YPZRWBKMTBYPTK-UHFFFAOYSA-N

Biochem/physiol Actions

Cell permeable: no
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Irritant (B)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiulan Shao et al.
Oncology letters, 21(6), 431-431 (2021-04-20)
Ovarian cancer is one of the leading causes of cancer-related death among women worldwide and accounts for 4% of all cancer cases in female patients. To date, ovarian cancer has the poorest prognosis among all types of gynecological cancer; thus

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service