25972-M
2-Deoxy-D-glucose
Non-metabolizable derivative of glucose that can be carried by the glucose carrier in human erythrocytes. An indirect inhibitor of hexokinase. Reported to reduce the incidence of doxorubicin-induced apoptosis.
Synonym(s):
2-Deoxy-D-glucose
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H12O5
CAS Number:
Molecular Weight:
164.16
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.79
Recommended Products
Quality Level
description
RTECS - MQ3325000
Merck USA index - 14, 2904
Assay
≥98% (HPLC)
form
powder
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
color
off-white to white
solubility
water: 5 mg/mL
storage temp.
2-8°C
SMILES string
O1C(CC(C(C1CO)O)O)O
InChI
1S/C6H12O5/c7-2-4-6(10)3(8)1-5(9)11-4/h3-10H,1-2H2
InChI key
PMMURAAUARKVCB-UHFFFAOYSA-N
General description
Non-metabolizable derivative of glucose that can be carried by the glucose carrier in human erythrocytes. An indirect inhibitor of hexokinase. Reported to reduce the incidence of doxorubicin-induced apoptosis.
Warning
Toxicity: Standard Handling (A)
Reconstitution
Following reconstitution, refrigerate (4°C). Stock solutions are stable for up to 6 months at 4°C.
Other Notes
Thakkar, N.S., and Potten, C.S. 1993. Cancer Res. 53, 2057.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service