Skip to Content
Merck
All Photos(1)

Key Documents

237911

Sigma-Aldrich

Creatine Phosphate dipotassium salt

≥98% (TLC), solid, high-energy phosphate compound, Calbiochem®

Synonym(s):

Creatine Phosphate, Dipotassium Salt, Phosphocreatine, 2K

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8K2N3O5P
CAS Number:
Molecular Weight:
287.29
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Creatine Phosphate, Dipotassium Salt, A high-energy phosphate compound. Transfer of phosphate group from creatine phosphate to ADP and from ATP to creatine form the basis of the creatine-creatine phosphate energy shuttle.

Quality Level

description

Merck USA index - 14, 7340

Assay

≥98% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

Related Categories

General description

A high-energy phosphate compound. Transfer of phosphate group from creatine phosphate to ADP and from ATP to creatine form the basis of the creatine-creatine phosphate energy shuttle.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Tokuda, H., et al. 1990. Eur. J. Biochem.192, 583.
Bessman, S.P., and Carpenter, C.L. 1985. Annu. Rev. Biochem.54, 831.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

515.3 °F

Flash Point(C)

268.5 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service