Skip to Content
Merck
All Photos(1)

Key Documents

219426

Sigma-Aldrich

Cathepsin L Inhibitor II

The Cathepsin L Inhibitor II controls the biological activity of Cathepsin L. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

Z-FY-CHO

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

off-white

solubility

ethanol: 50 mg/mL
DMSO: soluble

shipped in

ambient

storage temp.

−20°C

General description

A potent and selective inhibitor of Cathepsin L (Cat. No. 219402).

Biochem/physiol Actions

Cell permeable: yes
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

Z-Phe-Tyr-CHO

Other Notes

Woo, J.-T., et al. 1996. Eur. J. Pharmacol. 300, 131.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Stephan Quintin et al.
The Journal of biological chemistry, 299(7), 104912-104912 (2023-06-13)
α-synuclein (αS) is an abundant, neuronal protein that assembles into fibrillar pathological inclusions in a spectrum of neurodegenerative diseases that include Lewy body diseases (LBD) and Multiple System Atrophy (MSA). The cellular and regional distributions of pathological inclusions vary widely
Prashant Gaikwad et al.
microPublication biology, 2022 (2023-01-07)
Several studies have indicated a role for cathepsin L (CTSL) proteolytic activity in the nucleus under distinct cellular conditions, including during differentiation, senescence, and quiescence. Here we show that addition of CTSL inhibitors to a cell lysis buffer prevents the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service