Skip to Content
Merck
All Photos(1)

Key Documents

218750

Sigma-Aldrich

Caspase-3 Inhibitor III

The Caspase-3 Inhibitor III, also referenced under CAS 285570-60-7, controls the biological activity of Caspase-3. This small molecule/inhibitor is primarily used for Cancer applications.

Synonym(s):

Caspase-3 Inhibitor III, Ac-DEVD-CMK

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H31ClN4O11
CAS Number:
Molecular Weight:
550.94
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.54
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

lyophilized solid
powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

white to off-white

solubility

20% acetonitrile: 1 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

ClCC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)C)CC(=O)O)CCC(=O)O)C(C)C)CC(=O)O

InChI

1S/C21H31ClN4O11/c1-9(2)18(21(37)25-12(6-16(31)32)14(28)8-22)26-19(35)11(4-5-15(29)30)24-20(36)13(7-17(33)34)23-10(3)27/h9,11-13,18H,4-8H2,1-3H3,(H,23,27)(H,24,36)(H,25,37)(H,26,35)(H,29,30)(H,31,32)(H,33,34)/t11-,12-,13-,18-/m0/s1

InChI key

ATNOUPFYBMVFLD-RSLFNQERSA-N

General description

A potent cell-permeable and irreversible inhibitor of caspase-3, a member of the ICE-CED-3 family of cysteine proteases.
A potent, cell-permeable, and irreversible inhibitor of caspase-3 as well as caspase-6, caspase-7, caspase-8, and caspase-10.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
caspase-3, caspase-6, caspase-7, caspase-8, caspase-10
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Sequence

Ac-Asp-Glu-Val-Asp-CMK

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 4 months at -20°C.

Other Notes

Mocanu, M.M., et al. 2000. Br. J. Pharmacol.130, 197
Schrantz, N., et al. 1999. Cell Death Differ. 6, 445.
Thornberry, N.A., and Lazebnik, Y. 1998. Science 281, 1312.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service