Skip to Content
Merck
All Photos(1)

Key Documents

1.94501

Supelco

Glycerol

anhydrous, for analysis EMPLURA®

Synonym(s):

Glycerol, 1,2,3-Propanetriol, Glycerin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
92.09
Beilstein:
635685
MDL number:
UNSPSC Code:
12352104
grade:
anhydrous
bp:
182 °C/20 mmHg (lit.)
vapor pressure:
<1 mmHg ( 20 °C)

grade

anhydrous

vapor density

3.1 (vs air)

vapor pressure

<1 mmHg ( 20 °C)

product line

EMPLURA®

autoignition temp.

698 °F

refractive index

n20/D 1.474 (lit.)

bp

182 °C/20 mmHg (lit.)

mp

20 °C (lit.)

density

1.25 g/mL (lit.)

storage temp.

room temp

SMILES string

OCC(O)CO

InChI

1S/C3H8O3/c4-1-3(6)2-5/h3-6H,1-2H2

InChI key

PEDCQBHIVMGVHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

1.94501.0521: 500ML, Minimum order quantity: 20

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

390.2 °F - Pensky-Martens closed cup

Flash Point(C)

199 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Inorganics and solvents for basic and standard applications manufactured locally.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service