Skip to Content
Merck
All Photos(1)

Key Documents

1.46768

Millipore

Centrimide Nalidixic Agar Kit

Synonym(s):

CN Agar, Pseudomonas Centrimide Nalidixic Agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106217
NACRES:
NA.74

material

gridded white mixed cellulose esters (MCE) membrane

Quality Level

form

plate (ready-to-use)

shelf life

9 mo.

feature

ready-to-use

manufacturer/tradename

ReadyPlate

packaging

pkg of (150x55mm Plates and 150 membranes)

pore size

0.45 μm pore size

application(s)

filtration
food and beverages
microbiology

storage temp.

15-25°C

suitability

Pseudomonas aeruginosa

General description

Centrimide Nalidixic Agar Kit contains
ReadyPlate CN Agar plates of 55mm diameter is a selective and differential chromogenic culture medium. It is a standard medium in water testing for simultaneous isolation and enumeration of Pseudomonas aeruginosa from water samples. Gelatine peptone and Casein hydrolysate supply amino acids and other complex nitrogenous substances. In the presence of Cetrimide (a selective agent that acts as a quaternary ammonium cationic detergent), the growth of bacterial cells other than P. aeruginosa is inhibited due to the release of nitrogen and phosphorous. Nalidixic acid present in the supplement also improves the inhibition of the accompanying microbial flora. The amount of potassium sulphate and magnesium chloride enhances the pigment production of Pseudomonas aeruginosa.
EZ-Pak Filters are ready-to-use, sterile filters made of Mixed cellulose esters (MCE) with 0.45 μm pore size, white, gridded and 50mm diameter. These filters possess high recoveries.

Application

Centrimide Nalidixic Agar Kit contains 55mm plates with EZPAK 0.45 μm white gridded membrane for the simultaneous enumeration of Pseudomonas aeruginosa using the Membrane Filtration method in water samples

Legal Information

READYPLATE is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service