1.46029
HAYFLICK Agar
suitable for microbiology, For general mycoplasma detection, pkg of 60 mm plates (20 plates per box)
Synonym(s):
Mycoplasma Agar
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Agency
EP 2.6.7
USP
Quality Level
sterility
sterile; aseptically filled
form
plate
feature
ready-to-use
packaging
pkg of 60 mm plates (20 plates per box)
pH
7.8 ( in H2O)
application(s)
industrial qc
pharmaceutical
microbiology
storage temp.
2-8°C
suitability
mycoplasma
General description
HAYFLICK Agar is supplemented with horse serum to provide useable cholesterol and fatty acids. Deoxyribonucleic acid provides purine and pyrimidine bases, which cannot be synthesized by the mycoplasmas. Phenol red changes the color of the medium to yellow or purple, indicating the growth of mycoplasmas. Penicillin inhibits the growth of accompanying bacterial flora.
Application
HAYFLICK Agar is recommended for the general detection of mycoplasmas in pharmaceutical products, especially in bulk vaccines, cell banks, and virus cultures.
Features and Benefits
- Ready-to-use convenience as no further preparation steps are required.
- Pharmaceutical products such as Biopharmaceuticals, Vaccines, Cell cultures, and Virus cultures.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service