Skip to Content
Merck
All Photos(1)

Key Documents

1.18751

Supelco

COD Test, photometric

photometric, 0-150 mg/L (COD), Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
SE.11
Pricing and availability is not currently available.

Product Name

COD Cell Test for photometers of other manufacturers, photometric, 0-150 mg/L (COD), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 25 tests

analyte functional class(es)

chemical oxygen demand (COD)

measuring range

0-150 mg/L (COD)

technique(s)

photometry: suitable

compatibility

for use with photometers from other manufacturers

detection method

photometric (Oxidation with Chromosulfuric acid, determination as chromate)

storage temp.

15-25°C

General description

We offer Spectroquant® test kits for photometers of other manufactures. This COD cell test kit comes with pre-filled 15.4 mm round cells. They work seamlessly with other photometer brands holding this size of cuvettes. The tests work with the original programming data installed by the manufacturers according to the instrument’s operation manual.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A - STOT RE 2

Target Organs

Kidney

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service