Skip to Content
Merck
All Photos(1)

Key Documents

1.18308

Supelco

Dodecane-1-sulfonic acid sodium salt

for ion pair chromatography LiChropur

Synonym(s):

Dodecane-1-sulfonic acid sodium salt, Sodium laurylsulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H25NaO3S
CAS Number:
Molecular Weight:
272.38
MDL number:
UNSPSC Code:
12352107
EC Index Number:
219-200-6
NACRES:
NB.21
Pricing and availability is not currently available.

Quality Level

Assay

≥99% (acidimetric)

technique(s)

ion pair chromatography: suitable

transmittance

200 nm, ≥70%
220 nm, ≥90%
250 nm, ≥98%
(0.005 M; 1 cm in H2O)

pH

5.5-7.5 (20 °C, 100 g/L in H2O)

bulk density

330 kg/m3

storage temp.

15-25°C

SMILES string

[Na+].[S](=O)(=O)([O-])CCCCCCCCCCCC

InChI

1S/C12H26O3S.Na/c1-2-3-4-5-6-7-8-9-10-11-12-16(13,14)15;/h2-12H2,1H3,(H,13,14,15);/q;+1/p-1

InChI key

DAJSVUQLFFJUSX-UHFFFAOYSA-M

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service