Skip to Content
Merck
All Photos(1)

Key Documents

1.12156

Sigma-Aldrich

Europium(III) oxide

99+

Synonym(s):

Europium(III) oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Eu2O3
CAS Number:
Molecular Weight:
351.93
MDL number:
UNSPSC Code:
12352303
EC Index Number:
215-165-6
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

mp

2291 °C

density

7.42 g/cm3 at 20 °C

storage temp.

no temp limit

SMILES string

[Eu+3].[Eu+3].[O-2].[O-2].[O-2]

InChI

1S/2Eu.3O/q2*+3;3*-2

InChI key

AEBZCFFCDTZXHP-UHFFFAOYSA-N

Analysis Note

Assay (complexometric, calc. on the ignited basis): ≥ 99.0 %
Identity (ICP-OES): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service