Skip to Content
Merck
All Photos(2)

Key Documents

1.07747

Supelco

Silica gel 60 PF₂₅₄

for preparative thin layer chromatography

Synonym(s):

Silica gel

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41115711
EC Index Number:
231-545-4
NACRES:
NB.21

material

silica gel 60 matrix

Quality Level

form

powder

potency

>5000 mg/kg LD50, skin (Rabbit)

feature

binder Organic Polymer
fluorescent indicator

packaging

pkg of 2.5 kg

technique(s)

preparative TLC: suitable
thin layer chromatography (TLC): suitable

particle size

5-40 μm

pore size

60 Å medium pore diameter

pH

7 (20 °C, 100 g/L in H2O, slurry)

bp

2230 °C

mp

>1600 °C

bulk density

200‑800 kg/m3

storage temp.

no temp limit

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Standardized sorbents ensure reliable results every time. Our broad spectrum meets all kinds of needs. In TLC, silica gel 60 sorbent is considered the most versatile. Our assortment of silica gel 60 features different grades with particle sizes ranging from 5-40 µm: silica with a gypsum binder, silica with no foreign binder, and silica gel with a fluorescence indicator. We also offer high-quality aluminum oxide, cellulose microcrystalline, and kieselgur (also known as diatomaceous earth).

Application

  • A rapid preparative method for isolation of neutral and acidic glycosphingolipids by radial thin-layer chromatography.: This study outlines a swift and efficient preparative method using radial thin-layer chromatography to isolate both neutral and acidic glycosphingolipids, utilizing Silica gel 60 PF254. The methodology enhances the purity and yield of the glycosphingolipids isolated, contributing significantly to advancements in analytical chemistry techniques (Das KK et al., 1984).

Analysis Note

pH-value (10 % suspension): 6.5 - 7.5
Fe (Iron): ≤ 0.02 %
Particle size: 90% ≤55 µm
Application test: conforms
The data to the particle size are related to the pure Silica gel before additive addition.

related product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service