Skip to Content
Merck
All Photos(1)

Key Documents

1.03164

Supelco

Eriochrome cyanine R (C.I. 43820)

metal indicator, for analysis, reagent for aluminum

Synonym(s):

Eriochrome cyanine R (C.I. 43820)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H15Na3O9S
CAS Number:
Molecular Weight:
536.39
MDL number:
UNSPSC Code:
12352107
EC Index Number:
222-641-7
NACRES:
NB.21
Pricing and availability is not currently available.

Product Name

Eriochrome cyanine R (C.I. 43820), for analysis (reagent for aluminium)

Quality Level

potency

>2000 mg/kg LD50, oral (Rat)

loss

≤10% loss on drying, 110°C

pH

9.1 (24 °C, 10 g/L in H2O)

solubility

420 g/L

bulk density

590 kg/m3

λmax

434-440 nm (buffer pH 7.0)

storage temp.

2-30°C

SMILES string

[Na+].[S]1(=O)(=O)OC(c4c1cccc4)(c3cc(c(c(c3)C(=O)O)O)C)c2cc(c(c(c2)C(=O)O)O)C

InChI

1S/C23H18O9S.Na/c1-11-7-13(9-15(19(11)24)21(26)27)23(14-8-12(2)20(25)16(10-14)22(28)29)17-5-3-4-6-18(17)33(30,31)32-23;/h3-10,24-25H,1-2H3,(H,26,27)(H,28,29);/q;+1

InChI key

HJEGHSWGWSAZBS-UHFFFAOYSA-N

Application

  • Determination of Food Oxalates Using Silica-Titania Xerogel Modified with Eriochrome Cyanine R.: This research explores a novel method to determine oxalate concentrations in food using Eriochrome Cyanine R, highlighting the dye′s potential in enhancing analytical sensitivity and specificity (Morosanova et al., 2018).
  • Development of a Dispersive Liquid-Liquid Microextraction Method Combined with UV-Visible Spectrophotometry for Determination of Trace Aluminum(III) in Water, Wastewater, Food, Biological, and Pharmaceutical Samples.: This study demonstrates a new method utilizing Eriochrome Cyanine R for tracing aluminum levels across various samples, showcasing its utility in environmental and health-related chemical analyses (Birgani et al., 2017).
  • Direct, sensitive determination of trace amounts of dissolved ferric iron in natural water by light absorption ratio variation spectrometry.: Though not directly mentioning Eriochrome Cyanine R, this article focuses on related spectrometric techniques that can be adapted for dyes like Eriochrome Cyanine R to detect iron in natural water sources (Gao et al., 2005).
  • Determination of vanadium by solid-phase spectrophotometry after its preconcentration as an Eriochrome Cyanine R complex on a dextran-type exchanger.: This research introduces a method for vanadium determination using Eriochrome Cyanine R, underlining the dye′s role in the preconcentration and quantification of vanadium in complex matrices (Boudra et al., 1995).

Analysis Note

Identity (UV/VIS-Spectrum): passes test
Absorption maximum λmax. (buffer pH 7.0): 434 - 440 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.02 g/l; buffer pH 7.0; calc. on dried substance): 130 - 230
Loss on drying (110 °C): ≤ 10 %

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service