Skip to Content
Merck
All Photos(1)

Key Documents

1.00219

Supelco

Amidosulfuric acid

EMPLURA®

Synonym(s):

Amidosulfuric acid, Amidosulfonic acid, Sulphamic acid, Sulfamidic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
H3NO3S
CAS Number:
Molecular Weight:
97.09
MDL number:
UNSPSC Code:
12352301
EC Index Number:
226-218-8
NACRES:
NA.21
Pricing and availability is not currently available.

Quality Level

product line

EMPLURA®

Assay

≥98.5% (acidimetric)

form

solid

ign. residue

≤0.05%

pH

1.2 (10 g/L in H2O)

mp

205 °C (decomposition)

solubility

water: 213 g/L at 20 °C

density

2.13 g/cm3 at 20 °C

bulk density

600 kg/m3

storage temp.

no temp limit

SMILES string

[S](=O)(=O)([N+H3])[O-]

InChI

1S/H3NO3S/c1-5(2,3)4/h(H3,1,2,3,4)

InChI key

IIACRCGMVDHOTQ-UHFFFAOYSA-N

Related Categories

Analysis Note

Identity (IR-spectrum): passes test
Assay (acidimetric): ≥ 98.5 %
Residue on ignition: ≤ 0.05 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service