610014
Avanti
Filter Support
Avanti Research™ - A Croda Brand
Synonym(s):
Filter Support, 10mm Filter Supports (100/pk)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352211
NACRES:
NB.24
Recommended Products
manufacturer/tradename
Avanti Research™ - A Croda Brand
storage temp.
room temp
General description
Filter Support for Avanti′s Mini-Extruder.
Watch the video to learn more about the Avanti Research™ Mini-Extruder
Watch the video to learn more about the Avanti Research™ Mini-Extruder
Biochem/physiol Actions
This product is used with the extruder to provide extra support and improved flow rate. Filter support provides a flat surface to eliminate tearing or rupturing of the polycarbonate membrane. It is also used as a separator between membrane layers in serial stack filtration applications. The polyester filter support is binder free and has a thickness of 100 μm. The filter support disc is chemically inert.
Components
Whatman® drain disc 230300
Legal Information
Avanti Research is a trademark of Avanti Polar Lipids, LLC
Whatman is a registered trademark of Cytiva
also commonly purchased with this product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service