Skip to Content
Merck
All Photos(2)

Documents

250520P

Avanti

N-Dodecyl-Beta-D-Maltoside

85%, Avanti Polar Lipids

Synonym(s):

DDM; lauryl maltoside; dodecyl 4-O-α-D-glucopyranosyl-β-D-glucopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H46O11
CAS Number:
Molecular Weight:
510.62
UNSPSC Code:
12352211
NACRES:
NA.25

product name

N-Dodecyl-Beta-D-Maltoside (DDM, 85%), Avanti Research - A Croda Brand

Assay

>85% (contains <15% alpha isomer)

form

powder

packaging

pkg of 1 × 1 g (250520P-1g)

manufacturer/tradename

Avanti Research - A Croda Brand

application(s)

life science and biopharma

shipped in

dry ice

storage temp.

−20°C

InChI

1S/C24H46O11/c1-2-3-4-5-6-7-8-9-10-11-12-32-23-21(31)19(29)22(16(14-26)34-23)35-24-20(30)18(28)17(27)15(13-25)33-24/h15-31H,2-14H2,1H3/t15-,16-,17-,18+,19-,20-,21-,22-,23-,24-/m1/s1

InChI key

NLEBIOOXCVAHBD-QKMCSOCLSA-N

General description

This product is a more cost-effective alternative to Avandecrys® DDM that can be used for membrane protein extraction and purification. DDM is the most widely used detergent for the isolation and purification of GPCR′s. In addition, LacY likely favors a higher-energy intermediate periplasmic-open conformation in situ, but collapses to a lower-energy periplasmic-closed conformation in DDM after purification. It is interesting to note that the branched-chain maltoside detergents stabilize LacY in the membrane-embedded form

Packaging

20 mL Clear Glass Screw Cap Vial (250520P-1g)

Legal Information

Avandecrys is a registered trademark of Avanti Polar Lipids, Inc.
Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service