Skip to Content
Merck
All Photos(1)

Key Documents

Z699063

Total Count Broth with TTC

pack of 50 × 2 mL

Synonym(s):

agar and broth, agar broth, broth media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41111700
NACRES:
NB.79
Pricing and availability is not currently available.

packaging

pack of 50 × 2 mL

manufacturer/tradename

Maine Mfg 10496113

application(s)

food and beverages

storage temp.

2-8°C

General description

All bacteria develop on Total Count Media with indicator and produce a red color as a result of the precipitation of formazan following the reduction of 2,3,5-triphenyltetrazolium chloride (TTC) by bacteria.

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service