Z531154
Aldrich® coiled condenser
unjacketed, L 300 mm, joint: ST/NS 29/32
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Recommended Products
Joint
joint: ST/NS29/32
L
300 mm
Related Categories
General description
The coiled condenser circulates coolant through the coil. The jacketed version provides additional cooling capacity by allowing vapors to condense on the inner wall of the jacket.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)