Key Documents
Z341916
IKA® fixing clip attachment for HS/KS 260 basic and control shakers
AS 260.2 fixing clip attachment
Synonym(s):
flask clamps, holding clamps
About This Item
UNSPSC Code:
41103811
NACRES:
NB.73
packaging
set of 1
manufacturer/tradename
IKA 3115500
General description
Note: This attachment (without fixing clips) is for shaking flasks, Erlenmeyer flasks and bottles with a round cross-section.
Legal Information
IKA is a registered trademark of IKA-Werke GmbH & Co. KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)