Skip to Content
Merck
All Photos(1)

Key Documents

W400408

Sigma-Aldrich

2-(Methylthio)ethanol

≥99%

Synonym(s):

2-(Methylmercapto)ethanol, 2-Hydroxyethyl methyl sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SCH2CH2OH
CAS Number:
Molecular Weight:
92.16
FEMA Number:
4004
Beilstein:
1731081
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.179
NACRES:
NA.21

biological source

synthetic

Quality Level

Agency

meets purity specifications of JECFA

Assay

≥99%

refractive index

n20/D 1.4930 (lit.)

bp

169-171 °C (lit.)

density

1.06 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

meaty; sulfurous

SMILES string

CSCCO

InChI

1S/C3H8OS/c1-5-3-2-4/h4H,2-3H2,1H3

InChI key

WBBPRCNXBQTYLF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aaron B Cowley et al.
Biochemistry, 43(6), 1656-1666 (2004-02-11)
N-Acetylmicroperoxidase-8 (1) contains heme and residues 14-21 of horse mitochondrial cytochrome c (cyt c). The two thioether bonds linking protein to heme in cyt c are present in 1, and the native axial ligand His-18 remains coordinated to iron. As
Matthew C Mackenzie et al.
Biomacromolecules, 16(1), 236-245 (2014-12-18)
The field of RNA interference depends on the development of safe and efficient carriers for short interfering ribonucleic acid (siRNA) delivery. Conventional cationic monomers for siRNA delivery have utilized the nitrogen heteroatom to produce cationic charges. Here, we polymerized cationic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service