Skip to Content
Merck
All Photos(1)

Key Documents

W343900

Sigma-Aldrich

Myrtenol

≥95%, FG

Synonym(s):

(6,6-dimethyl-4-bicyclo[3.1.1]hept-3-enyl)methanol, 2-pinen-10-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
FEMA Number:
3439
EC Number:
Council of Europe no.:
10285
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.091
NACRES:
NA.21
Organoleptic:
fruity; herbaceous; minty
grade:
FG
Kosher
biological source:
synthetic
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

FG
Kosher

reg. compliance

EU Regulation 1334/2008 & 872/2012

Assay

≥95%

optical activity

[α]20/D −51°, neat

refractive index

n20/D 1.496 (lit.)

bp

221-222 °C (lit.)

density

0.954 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fruity; herbaceous; minty

SMILES string

CC1(C)C2CC=C(CO)C1C2

InChI

1S/C10H16O/c1-10(2)8-4-3-7(6-11)9(10)5-8/h3,8-9,11H,4-6H2,1-2H3/t8-,9-/m0/s1

InChI key

RXBQNMWIQKOSCS-IUCAKERBSA-N

Application


  • Combined antimicrobial action of streptomycin and terpenes against atypical mycobacteria isolated from fish.: Research demonstrates the synergistic effects of myrtenol with streptomycin in combating atypical mycobacteria, suggesting a potential for enhancing antibiotic treatments in aquaculture (Guz et al., 2024).


Biochem/physiol Actions

Taste at 50 ppm

Other Notes

Natural occurrence: Bilberry, buchu oil, hop oil, myrtle oil, pepper, raspberry, scotch spearment, strawberry, Virginia tobacco.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

192.2 °F - closed cup

Flash Point(C)

89 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mosciano, G.
Perfumer & Flavorist, 20, 52-52 (2005)
Organoleptic Characteristics of Flavor Materials
Mosciano, G.
Perfumer & Flavorist, 30, 54-54 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service