SYNC364625C
Synthware™ chromatography column with PTFE stopcock and coarse frit
L 254 mm
Synonym(s):
Synthware C364625C, glass chromatograpy column
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Recommended Products
grade
for chromatography
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C364625C
L
254 mm
Looking for similar products? Visit Product Comparison Guide
General description
With reinforced beaded top, coarse porosity fritted disc to support column packing. All Synthware columns are heavy wall designed to increase safety.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service