SYNA274100
Synthware™ anti-climb adapter
100 mL, top joint: ST/NS 24/40, bottom joint: ST/NS 24/40
Synonym(s):
Synthware A274100, bump trap adapter, bump trap
All Photos(1)
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
Recommended Products
Joint
bottom joint: ST/NS24/40
joint: ST/NS top24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A274100
capacity
100 mL
General description
Trap provides expanded volume for reactions that have a tendency to foam excessively. The inner vapor tube is sealed to the top of the The inner vapor tube is sealed to the top of thetrap to prevent entry into the condenser.
Legal Information
Synthware is a trademark of Kemtech America Inc.