CLS8160100
Pyrex® graduated oil centrifuge tube
capacity 100.0 mL, O.D. × L 38 mm × 200 mm
Synonym(s):
Corning® 8160
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121703
NACRES:
NB.43
Recommended Products
material
borosilicate glass
conical bottom tube
sterility
non-sterile
feature
cap: no
packaging
case of 12
pack of 12
manufacturer/tradename
Corning 8160-100
O.D. × L
38 mm × 200 mm
capacity
100.0 mL
color
clear
Related Categories
General description
In accordance with standard methods of testing for water and sediment in petroleum products. White graduations upward to 0.5 mL in divisions of 0.05 mL; from 0.5 mL to 2 mL in divisions of 0.1 mL; from 2 mL to 3 mL in divisions of 0.2 mL; from 3 mL to 5 mL in divisions of 0.5 mL; from 5 mL to 10 mL in divisions of 1 mL; from 10 mL to 25 mL in divisions of 5 mL and at 50, 75 and 100 mL.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service