CLS812012
Pyrex® centrifuge tubes heavy-duty with beaded rim
capacity 12 mL, ungraduated
Synonym(s):
Corning® 8120
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121703
NACRES:
NB.43
Recommended Products
material
borosilicate glass
conical bottom tube
sterility
non-sterile
packaging
case of 12
pack of 12
manufacturer/tradename
Corning 8120-12
O.D. × L
17 mm × 120 mm
capacity
12 mL
color
clear
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Always use with correct sleeve or holder. Increased wall thickness for greater mechanical strength. Formed from borosilicate glass. Stress that glass can withstand is greatly decreased by surface flaws such as scratches, chips, and cracks.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service