Key Documents
CLS2480200
Pyrex® Allihn condenser, with male and female joints
jacket L 200 mm, joint: ST/NS 19/38
Synonym(s):
laboratory condenser
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Joint
joint: ST/NS19/38
manufacturer/tradename
Corning 2480-200
jacket L
200 mm
General description
Suitable for use in vacuum distillation. With drip-tip
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)