BR37268
BRAND® volumetric flask BLAUBRAND® class A DE-M marking
capacity 20 mL, neck joint: ST/NS 12/21, glass stopper
Synonym(s):
BLAUBRAND®, Volumetric flasks
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass
glass stopper
Joint
joint: ST/NS neck12/21
feature
class A DE-M marking. DIN EN ISO 1042. Calibrated to contain (TC, In).
packaging
pack of 2 ea
manufacturer/tradename
BRAND 37268
capacity
20 mL
Looking for similar products? Visit Product Comparison Guide
General description
The BRAND® volumetric flask BLAUBRAND® class A with DE-M marking is made from high-quality Boro 3.3 glass and complies with DIN EN ISO 1042 standards. It is calibrated to contain (TC, In), ensuring accurate liquid measurement for various laboratory applications. Each volumetric flask is delivered with a batch number and batch certificate included in the original packaging, providing quality assurance and traceability. This reliable volumetric flask is an essential tool for precise analytical work in any laboratory setting.
Legal Information
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service