BR114130
BRAND® culture tube, rimless
L × W 18 mm × 180 mm, thickness 0.8 mm
Synonym(s):
Culture tubes, Tubes
About This Item
Recommended Products
material
round bottom
screw top cap
soda-lime glass tube
straight neck
transparent tube
sterility
non-sterile
feature
RCF 900 × g
neck thread push fit
packaging
pack of 121 ea
manufacturer/tradename
BRAND 114130
L × W
18 mm × 180 mm
capacity
18 mL
30 mL
thickness
0.8 mm
General description
Features and Benefits
- Excellent sample visibility
- Good chemical resistance
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)