932124
SF3-TRZ
≥99% (HPLC)
Synonym(s):
2-(9,9′-Spirobi[fluoren]-3-yl)-4,6-diphenyl-1,3,5-triazine
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
grade
sublimed grade
Quality Level
Assay
≥99% (HPLC)
description
3.44 eV (EG: HOMO-LUMO gap)
solubility
toluene: soluble
Orbital energy
HOMO -6.54 eV
LUMO -3.10 eV
Application
SF3-TRZ can also be utilized in devices such as organic photovoltaic (OPV) devices and organic field-effect transistors (OFETs). In OPV devices, it can serve as an electron transport or electron-blocking material, assisting in efficient charge extraction and collection. In OFETs, SF3-TRZ can act as an electron-transport layer, enabling the smooth flow of electrons in the device. It can be used as an electron-transport layer in organic light-emitting diodes (OLEDs), facilitating the efficient injection and transport of negative charges (electrons) from the cathode to the emissive layer.
Use as TADF Host for Green TADF OLED, EQE of 20.6%.
µ0 (hole) : ~9 × 10−9 cm2 V−1 s−1
µ0(electron): ~4 × 10−5 cm2 V−1 s−1
µ0 (hole) : ~9 × 10−9 cm2 V−1 s−1
µ0(electron): ~4 × 10−5 cm2 V−1 s−1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Nature communications, 8(1), 2250-2250 (2017-12-23)
Organic light-emitting diodes have become a mainstream display technology because of their desirable features. Third-generation electroluminescent devices that emit light through a mechanism called thermally activated delayed fluorescence are currently garnering much attention. However, unsatisfactory device stability is still an
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service