Skip to Content
Merck
All Photos(3)

Key Documents

900563

Sigma-Aldrich

Poly(3-hexylthiophene-2,5-diyl)

regioregular, average Mw 20,000-45,000

Synonym(s):

P3HT

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C10H14S)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

description

Metal purity: Mg: ≤100 ppm
Ni: ≤100 ppm

Regioregularity: ≥90%

Quality Level

form

solid

mol wt

average Mw 20,000-45,000

SMILES string

[s]1c(c(cc1C)CCCCCC)C

InChI

1S/C12H20S/c1-4-5-6-7-8-12-9-10(2)13-11(12)3/h9H,4-8H2,1-3H3

InChI key

DUFPJSOXRHVDOV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(3-hexylthiophene-2,5-diyl) (P3HT) is a polythiophene-based low bandgap semiconducting polymer. It is widely used as anelectron donor material in organic solar cells, OLEDs, and OFETs due to itschemical and thermal stability, controlled synthetic method, goodsemiconducting properties, simple structure, and widecommercial availability. The degree of regioregularity of P3HT has a stronginfluence on its ability to form highly ordered domains.

Application

Poly(3-hexylthiophene-2,5-diyl) can be used as a:

  • Active semiconductor to prepare organic field emission transistors. In regioregular P3HT the microcrystalline domains are well constructed by phase-segregated layers of insulating alkali side chains and pi-pi interchain stacking. This leads to high charge mobility and enhances the performance of OFETs.
  • Precursor to synthesize P3HT-PCBM( [6,6]-phenyl C61-butyric acid methyl ester) blend thin film. The highcrystallinity of P3HT thin film increases the efficiency of organicphotovoltaic cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Morongwa E Ramoroka et al.
Polymers, 13(1) (2020-12-31)
This report focuses on the synthesis of novel 2,3,4,5-tetrathienylthiophene-co-poly(3-hexylthiophene-2,5-diyl) (TTT-co-P3HT) as a donor material for organic solar cells (OSCs). The properties of the synthesized TTT-co-P3HT were compared with those of poly(3-hexylthiophene-2,5-diyl (P3HT). The structure of TTT-co-P3HT was studied using nuclear

Articles

Professor Shinar highlights low-cost, disposable sensor configurations in organic and hybrid electronics for healthcare applications.

Professor Shinar highlights low-cost, disposable sensor configurations in organic and hybrid electronics for healthcare applications.

Professor Shinar highlights low-cost, disposable sensor configurations in organic and hybrid electronics for healthcare applications.

Small molecular weight organic semiconductors are promising for flexible transistor applications in next-gen soft electronics.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service