80723
(Piperidinomethyl)polystyrene
extent of labeling: ~3.5 mmol/g base loading
Synonym(s):
1-Benzylpiperidine polymer-bound, Copolymer of styrene and divinylbenzene, piperidinomethylated, Piperidine, polymer-bound, Polystyrene crosslinked with divinylbenzene, piperidinomethylated
About This Item
Recommended Products
form
solid
reaction suitability
reaction type: solution phase peptide synthesis
reactivity: proton reactive
extent of labeling
~3.5 mmol/g base loading
impurities
≤0.2 mmol/g Cl (resin)
matrix
crosslinked with 2% DVB
particle size
200-400 mesh
Application
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service