Skip to Content
Merck
All Photos(1)

Documents

769878

Sigma-Aldrich

Resomer® LG 857 S, Poly(L-lactide-co-glycolide)

ester terminated, lactide:glycolide 85:15

Synonym(s):

Resomer® LG 857 S, PLGA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO[C3H4O2]n[C2H2O2]mCH3
CAS Number:
UNSPSC Code:
12162002
NACRES:
NA.23

Quality Level

form

solid

feed ratio

lactide:glycolide 85:15

viscosity

5.0-7.0 dL/g

mp

159-166 °C

transition temp

Tg 53-63 °C

storage temp.

2-8°C

Application

Biodegradable, biocompatible, and bioresorbable copolymer composed of L-Lactide and Glycolide. This semi-crystalline material has been used in the fabrication of research medical devices and research tissue engineering solutions, such as orthopedic or soft tissue fixation devices. Degradation of this materials has been thoroughly studied and has been shown to be safely resorbed by the body after implantation. Modification of molecular weight and polymer composition allows for control of the degradation rate and mechanical stability of the polymer.

Legal Information

RESOMER is a registered trademark of Evonik Rohm GmbH

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service