Skip to Content
Merck
All Photos(1)

Key Documents

752797

Sigma-Aldrich

[(R,R)-Teth-TsDpen RuCl]

Synonym(s):

chloro[N-[(1R,2R)-1,2-diphenyl-2-[[3-(η6-phenyl)propyl]amino-κN]ethyl]-4-methylbenzenesulfonamidato-κN]- Ruthenium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H31ClN2O2RuS
CAS Number:
Molecular Weight:
620.17
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

form

solid

optical activity

[α]22/D -302°, c = 0.5% in chloroform

reaction suitability

core: ruthenium
reagent type: catalyst

mp

260-268 °C

storage temp.

2-8°C

SMILES string

Cc1ccc(cc1)S(=O)(=O)N2[C@@H]([C@H]([N@H](CCCc3ccccc3)[Ru]2Cl)c4ccccc4)c5ccccc5

InChI

1S/C30H31N2O2S.ClH.Ru/c1-24-19-21-28(22-20-24)35(33,34)32-30(27-17-9-4-10-18-27)29(26-15-7-3-8-16-26)31-23-11-14-25-12-5-2-6-13-25;;/h2-10,12-13,15-22,29-31H,11,14,23H2,1H3;1H;/q-1;;+2/p-1/t29-,30-;;/m1../s1

InChI key

MDABGVLQRDDWLY-SEILFYAJSA-M

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service