Skip to Content
Merck
All Photos(1)

Documents

699373

Sigma-Aldrich

1,2-Dichloropropane-d6

98 atom % D, 98% (CP)

Synonym(s):

Propylene dichloride-d6

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3D6Cl2
Molecular Weight:
119.02
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

isotopic purity

98 atom % D

Assay

98% (CP)

form

liquid

bp

95-96 °C (lit.)

mp

-100 °C (lit.)

density

1.216 g/mL at 25 °C

mass shift

M+6

SMILES string

[2H]C([2H])([2H])C([2H])(Cl)C([2H])([2H])Cl

InChI

1S/C3H6Cl2/c1-3(5)2-4/h3H,2H2,1H3/i1D3,2D2,3D

InChI key

KNKRKFALVUDBJE-LIDOUZCJSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

59.0 °F - closed cup - Based on data from similar materials

Flash Point(C)

15.0 °C - closed cup - Based on data from similar materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service