Skip to Content
Merck
All Photos(3)

Key Documents

687073

Sigma-Aldrich

4′-(4-Chlorophenyl)-2, 2′:6′, 2″-terpyridine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H14ClN3
CAS Number:
Molecular Weight:
343.81
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

solid

mp

168-172 °C

SMILES string

Clc1ccc(cc1)-c2cc(nc(c2)-c3ccccn3)-c4ccccn4

InChI

1S/C21H14ClN3/c22-17-9-7-15(8-10-17)16-13-20(18-5-1-3-11-23-18)25-21(14-16)19-6-2-4-12-24-19/h1-14H

InChI key

UPOMZZQMFGCHGB-UHFFFAOYSA-N

Application

Terpyridine ligands (building block) for making organometallic complexes used in solar cells.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 4

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Winter, A.; van den Berg, Antje M. J.; et al.
Synthesis, 17, 2873-2878 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service