Skip to Content
Merck
All Photos(1)

Key Documents

674745

Sigma-Aldrich

(R)-(–)-3,3′-Bis(triphenylsilyl)-1,1′-binaphthyl-2,2′-diyl hydrogenphosphate

95%

Synonym(s):

(11bR)-4-Hydroxy-2,6-bis(triphenylsilyl)dinaphtho-[2,1-d:1′,2′f]-1,3,2-dioxaphosphepin 4-oxide, MacMillan TiPSY catalyst

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C56H41O4PSi2
CAS Number:
Molecular Weight:
865.07
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

solid

optical activity

[α]22/D -197°, c = 1 in chloroform

mp

329-335 °C

SMILES string

OP1(=O)Oc2c(cc3ccccc3c2-c4c(O1)c(cc5ccccc45)[Si](c6ccccc6)(c7ccccc7)c8ccccc8)[Si](c9ccccc9)(c%10ccccc%10)c%11ccccc%11

InChI

1S/C56H41O4PSi2/c57-61(58)59-55-51(62(43-25-7-1-8-26-43,44-27-9-2-10-28-44)45-29-11-3-12-30-45)39-41-23-19-21-37-49(41)53(55)54-50-38-22-20-24-42(50)40-52(56(54)60-61)63(46-31-13-4-14-32-46,47-33-15-5-16-34-47)48-35-17-6-18-36-48/h1-40H,(H,57,58)

InChI key

BDQOCXQVIFQJRK-UHFFFAOYSA-N

Application

Organocatalyst for the first enatioselective organocatalytic reductive amination reaction.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Ian Storer et al.
Journal of the American Chemical Society, 128(1), 84-86 (2006-01-05)
The first enantioselective organocatalytic reductive amination reaction has been accomplished. The development of a new chiral phosphoric acid catalyst has provided a convenient strategy for the enantioselective construction of protected primary amines and provided a highly stereoselective method for the

Articles

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service