672882
3,4-Difluorophenylacetylene
90%
Synonym(s):
4-Ethynyl-1,2-difluorobenzene
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C8H4F2
CAS Number:
Molecular Weight:
138.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
90%
refractive index
n20/D 1.4902
density
1.144 g/mL at 25 °C
functional group
fluoro
SMILES string
Fc1ccc(cc1F)C#C
InChI
1S/C8H4F2/c1-2-6-3-4-7(9)8(10)5-6/h1,3-5H
InChI key
XFPAOXIWRDDQGG-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
45.0 °F - closed cup
Flash Point(C)
7.22 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service