Skip to Content
Merck
All Photos(1)

Key Documents

666572

Sigma-Aldrich

Dicyclohexylphosphine

97%

Synonym(s):

Cy2PH

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H11)2PH
CAS Number:
Molecular Weight:
198.28
EC Number:
MDL number:
UNSPSC Code:
12350000
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

reaction suitability

reagent type: ligand
reaction type: Suzuki-Miyaura Coupling

refractive index

n20/D 1.516

bp

129 °C/8 mmHg

functional group

phosphine

storage temp.

20-25°C

SMILES string

C1CCC(CC1)PC2CCCCC2

InChI

1S/C12H23P/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12/h11-13H,1-10H2

InChI key

HDULBKVLSJEMGN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Pyr. Liq. 1

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service