Skip to Content
Merck
All Photos(1)

Key Documents

657670

Sigma-Aldrich

Poly(2-dimethylamino)ethyl methacrylate) methyl chloride quaternary salt

Synonym(s):

MADQUAT, Poly(2-(trimethylamino)ethyl methacrylate), Poly(2-methacryloxyethyltrimethylammonium chloride)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C9H18ClNO2)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23
Pricing and availability is not currently available.

form

solid

Quality Level

solubility

H2O: soluble
methanol: soluble

SMILES string

[Cl-].[N+](CCOC(=O)C(=C)C)(C)(C)C

InChI

1S/C9H18NO2.ClH/c1-8(2)9(11)12-7-6-10(3,4)5;/h1,6-7H2,2-5H3;1H/q+1;/p-1

InChI key

RRHXZLALVWBDKH-UHFFFAOYSA-M

Physical properties

Cationic polyelectrolyte[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Direct Synthesis of Well-Defined Quaternized Homopolymers and Diblock Copolymers via ATRP in Protic Media.
Li Y, et al.
Macromolecules, 36, 8268-8268 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service