Skip to Content
Merck
All Photos(2)

Key Documents

631264

Sigma-Aldrich

3-Iodo-4-methylaniline

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
IC6H3(CH3)NH2
CAS Number:
Molecular Weight:
233.05
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

form

solid

mp

37-41 °C (lit.)

functional group

iodo

SMILES string

Cc1ccc(N)cc1I

InChI

1S/C7H8IN/c1-5-2-3-6(9)4-7(5)8/h2-4H,9H2,1H3

InChI key

RRUDMHNAMZFNEK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Iodo-4-methylaniline can undergo reaction with paraformaldehyde to give the corresponding dihalo-substituted analogs of Troger′s base.[1] It can be also be used in the synthesis of BIRB 796 (1-(5-tert-butyl-2-p-tolyl-2H-pyrazol-3-yl)-3-[4-(2-morpholin-4-yl-ethoxy)naphthalen-1-yl]urea), a promising agent for the treatment of inflammatory diseases.[2]

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Synthesis of Dihalo-Substituted Analogues of Troger?s Base from ortho-and meta-Substituted Anilines"
Hansson A, et al.
European Journal of Organic Chemistry, 2003(16), 3179-3188 (2003)
"Synthesis of deuterium, tritium, and carbon-14 labeled BIRB 796, a p38 MAP kinase inhibitor"
Latli B
Journal of Labelled Compounds & Radiopharmaceuticals, 47(12), 847-856 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service