609811
2-Propanol-17O
20 atom % 17O
Synonym(s):
Isopropyl alcohol-17O
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
isotopic purity
20 atom % 17O
mol wt
60.30 by atom % calculation (20 atom % 17O)
bp
82 °C
density
0.798 g/mL at 25 °C
mass shift
M+1
SMILES string
CC(C)[17OH]
InChI
1S/C3H8O/c1-3(2)4/h3-4H,1-2H3/i4+1
InChI key
KFZMGEQAYNKOFK-AZXPZELESA-N
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
53.6 °F
Flash Point(C)
12 °C
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service