Skip to Content
Merck
All Photos(1)

Key Documents

527009

Sigma-Aldrich

(4-Phenylthiophenyl)diphenylsulfonium triflate

Synonym(s):

(4-Phenylthiophenyl)diphenylsulfonium trifluoromethanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H19F3O3S3
CAS Number:
Molecular Weight:
520.61
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

mp

81-85 °C (lit.)

Quality Level

λmax

298 nm

SMILES string

[O-]S(=O)(=O)C(F)(F)F.S(c1ccccc1)c2ccc(cc2)[S+](c3ccccc3)c4ccccc4

InChI

1S/C24H19S2.CHF3O3S/c1-4-10-20(11-5-1)25-21-16-18-24(19-17-21)26(22-12-6-2-7-13-22)23-14-8-3-9-15-23;2-1(3,4)8(5,6)7/h1-19H;(H,5,6,7)/q+1;/p-1

InChI key

WEYUQUMMYNRIPP-UHFFFAOYSA-M

Application

Cationic photoinitiator. Photoacid generator.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Baptiste Monney et al.
Journal of materials chemistry. B, 8(30), 6357-6365 (2020-06-20)
Neural implants that are based on mechanically adaptive polymers (MAPs) and soften upon insertion into the body have previously been demonstrated to elicit a reduced chronic tissue response than more rigid devices fabricated from silicon or metals, but their processability

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service