Skip to Content
Merck
All Photos(1)

Key Documents

490946

Sigma-Aldrich

Trimethylamine-15N

98 atom % 15N

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)315N
CAS Number:
Molecular Weight:
60.10
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

vapor density

2.09 (vs air)

vapor pressure

13.3 psi ( 0 °C)

isotopic purity

98 atom % 15N

autoignition temp.

374 °F

bp

3-4 °C (lit.)

mp

−117 °C (lit.)

density

0.647 g/mL at 25 °C (lit.)

mass shift

M+1

SMILES string

C[15N](C)C

InChI

1S/C3H9N/c1-4(2)3/h1-3H3/i4+1

InChI key

GETQZCLCWQTVFV-AZXPZELESA-N

Related Categories

Packaging

Standard 1 g quantitiy is packaged in a 25 mL Sure/Pac cylinder with stainless steel 1/4" male NPT valve. Nominal gas pressure at 21ºC is 13 psig for 1 g. This pressure is slightly above atmospheric pressure. Care must be taken when extracting this product from the cylinder.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Recommended products

Stainless steel hose adapter Z146838 or stainless steel body mini gas regulator Z513547 is recommended.

Legal Information

Sure/Pac is a trademark of Sigma-Aldrich Co. LLC

hose barb

Product No.
Description
Pricing

regulator

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Gas 1A - Press. Gas Liquefied gas - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

2A - Gases

WGK

WGK 1

Flash Point(F)

19.9 °F

Flash Point(C)

-6.7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service