Skip to Content
Merck
All Photos(1)

Key Documents

489417

Sigma-Aldrich

Formaldehyde-13C solution

20 wt. % in H2O, 99 atom % 13C

Synonym(s):

13C Labeled formaldehyde solution, 13C Labeled formalin solution, Formalin-13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H13CHO
CAS Number:
Molecular Weight:
31.02
MDL number:
UNSPSC Code:
12352501
PubChem Substance ID:
NACRES:
NA.12

vapor pressure

52 mmHg ( 37 °C)

Quality Level

isotopic purity

99 atom % 13C

form

liquid

concentration

20 wt. % in H2O

technique(s)

protein expression: suitable

mass shift

M+1

storage temp.

room temp

SMILES string

[H][13C]([H])=O

InChI

1S/CH2O/c1-2/h1H2/i1+1

InChI key

WSFSSNUMVMOOMR-OUBTZVSYSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service