484202
Di(propylene glycol) methyl ether acetate, mixture of isomers
≥98%
Synonym(s):
2-(2-methoxypropoxy)propyl acetate, DOWANOL® DPMA
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3CO2C3H6OC3H6OCH3
CAS Number:
Molecular Weight:
190.24
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23
Recommended Products
Quality Level
Assay
≥98%
refractive index
n20/D 1.417 (lit.)
bp
200 °C (lit.)
density
0.97 g/mL at 25 °C (lit.)
SMILES string
O(C(OC(=O)C)CC)CCCOC
InChI
1S/C9H18O4/c1-4-9(13-8(2)10)12-7-5-6-11-3/h9H,4-7H2,1-3H3
InChI key
USDXBNSCKZJDLC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Di(propylene glycol) methyl ether acetate can be used as a solvent in the preparation of electrically conductive adhesives.
It can also be used as an additive in solvent ink, for the preparation of wax-printed paper-based devices.
It can also be used as an additive in solvent ink, for the preparation of wax-printed paper-based devices.
Legal Information
Product of Dow Chemical
DOWANOL is a registered trademark of The Dow Chemical Company or an affiliated company of Dow
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
186.8 °F - closed cup
Flash Point(C)
86 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service