Skip to Content
Merck
All Photos(2)

Key Documents

437433

Sigma-Aldrich

Di(ethylene glycol) diacrylate

technical grade, 75%

Synonym(s):

2,2′-Oxydiethyl diacrylate, Diethylene glycol diacrylate, Polyethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(H2C=CHCO2CH2CH2)2O
CAS Number:
Molecular Weight:
214.22
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

grade

technical grade

Assay

75%

form

liquid

contains

60-100 ppm HQ as inhibitor
90-150 ppm MEHQ as inhibitor

reaction suitability

reagent type: cross-linking reagent
reaction type: Polymerization Reactions

refractive index

n20/D 1.463 (lit.)

bp

162 °C (lit.)

density

1.118 g/mL at 25 °C (lit.)

Ω-end

acrylate

α-end

acrylate

polymer architecture

shape: linear
functionality: homobifunctional

SMILES string

C=CC(=O)OCCOCCOC(=O)C=C

InChI

1S/C10H14O5/c1-3-9(11)14-7-5-13-6-8-15-10(12)4-2/h3-4H,1-2,5-8H2

InChI key

LEJBBGNFPAFPKQ-UHFFFAOYSA-N

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service