Skip to Content
Merck
All Photos(1)

Key Documents

334413

Sigma-Aldrich

Molecular sieves

powder, Catalyst support, ammonium Y zeolite

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
23201100
eCl@ss:
32110204
NACRES:
SB.52
Pricing and availability is not currently available.

form

powder

particle size

100 mesh (>90%)

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thermal decomposition of ammonium Y zeolite.
Ward JW.
J. Catal., 18(3), 348-351 (1970)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service