Skip to Content
Merck
All Photos(3)

Key Documents

327697

Sigma-Aldrich

Tris[2-(diphenylphosphino)ethyl]phosphine

greener alternative

97%

Synonym(s):

Tetraphos-Li

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(C6H5)2PCH2CH2]3P
CAS Number:
Molecular Weight:
670.68
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

134-135 °C (lit.)

functional group

phosphine

greener alternative category

SMILES string

C(CP(c1ccccc1)c2ccccc2)P(CCP(c3ccccc3)c4ccccc4)CCP(c5ccccc5)c6ccccc6

InChI

1S/C42H42P4/c1-7-19-37(20-8-1)44(38-21-9-2-10-22-38)34-31-43(32-35-45(39-23-11-3-12-24-39)40-25-13-4-14-26-40)33-36-46(41-27-15-5-16-28-41)42-29-17-6-18-30-42/h1-30H,31-36H2

InChI key

TVLNGWSWPKIYAO-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

Ligand used in iron-catalyzed, greener reduction of aldehydes to alcohols under transfer hydrogenation conditions.

Fast and selective iron-catalyzed transfer hydrogenations of aldehydes

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service