Skip to Content
Merck
All Photos(2)

Key Documents

256269

Sigma-Aldrich

(S,S)-(−)-Hydrobenzoin

99%, optical purity ee: 99% (GLC)

Synonym(s):

(S,S)-(−)-1,2-Diphenyl-1,2-ethanediol, (S,S)-1,2-Diphenylethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(OH)CH(OH)C6H5
CAS Number:
Molecular Weight:
214.26
Beilstein:
2330888
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

optical activity

[α]24/D −94°, c = 2.5 in ethanol

optical purity

ee: 99% (GLC)

mp

148-150 °C (lit.)

functional group

hydroxyl
phenyl

SMILES string

O[C@H]([C@@H](O)c1ccccc1)c2ccccc2

InChI

1S/C14H14O2/c15-13(11-7-3-1-4-8-11)14(16)12-9-5-2-6-10-12/h1-10,13-16H/t13-,14-/m0/s1

InChI key

IHPDTPWNFBQHEB-KBPBESRZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chiral reagent.

Application

(S,S)-(-)-Hydrobenzoin may also be used to prepare (1S,2S,1′S)- and (1S,2S,1′R)-2-(cyclohex-2′-enyloxy)-1,2-diphenylethanol, which are intermediates to prepare enantiopure cyclohexitols. The (S,S)-(-)-hydrobenzoin/Ca complex may be used to catalyze the direct asymmetric aldol reaction of acetophenone and pivalaldehyde to form (R)-3-hydroxy-4,4-dimethyl-1-phenylpentan-1-one.
C2 symmetric chiral diol with versatile applications as a chiral auxiliary, building block, and chiral ligand.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catalytic asymmetric aldol reaction of ketones and aldehydes using chiral calcium alkoxides.
Tetrahedron Letters, 42(28), 4669-4671 (2001)
Synthesis of enantiopure cyclitols from (?)-3-bromocyclohexene mediated by intramolecular oxyselenenylation employing (S, S)-hydrobenzoin and (S)-mandelic acid as chiral sources.
Lee YJ, et al.
Tetrahedron, 61(8), 1987-2001 (2005)
Burk, M.J. et al.
Tetrahedron Asymmetry, 2, 569-569 (1991)
Ho, O.C. et al.
Organometallics, 14, 2855-2855 (1995)
Pichon, M. Figadere, B.
Tetrahedron Asymmetry, 7, 927-927 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service