Merck
Preclinical Evaluation of a Lentiviral Vector for Huntingtin Silencing.
Molecular therapy. Methods & clinical development (2017-06-13)
Karine Cambon, Virginie Zimmer, Sylvain Martineau, Marie-Claude Gaillard, Margot Jarrige, Aurore Bugi, Jana Miniarikova, Maria Rey, Raymonde Hassig, Noelle Dufour, Gwenaelle Auregan, Philippe Hantraye, Anselme L Perrier, Nicole Déglon
RÉSUMÉ
Huntington's disease (HD) is an autosomal dominant neurodegenerative disorder resulting from a polyglutamine expansion in the huntingtin (HTT) protein. There is currently no cure for this disease, but recent studies suggest that RNAi to downregulate the expression of both normal and mutant HTT is a promising therapeutic approach. We previously developed a small hairpin RNA (shRNA), vectorized in an HIV-1-derived lentiviral vector (LV), that reduced pathology in an HD rodent model. Here, we modified this vector for preclinical development by using a tat-independent third-generation LV (pCCL) backbone and removing the original reporter genes. We demonstrate that this novel vector efficiently downregulated
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Valpromide, ≥97% (NMR)